accessibility menu, dialog, popup

UserName

H3COSiOH3COCH3

CAS RN 143487-47-2

IUPAC Name: cyclopentyltrimethoxysilane
Synonyms: cyclopentyltrimethoxysilane
Molecular Weight (g/mol): 190.31
Molecular Formula: C8H18O3Si
InChi Key: YRMPTIHEUZLTDO-UHFFFAOYSA-N
SMILES: CO[Si](OC)(OC)C1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results