missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentylmagnesium bromide, 2.0M solution in diethyl ether, AcroSeal™

Catalog No. p-4089912
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC427451000 100 mL AcroSeal™ Glass Bottle
AC427458000 800 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC427451000 Supplier Thermo Scientific Chemicals Supplier No. 427451000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 33240-34-5 | C5H9BrMg | 173.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33240-34-5 , 60-29-7
Molecular Formula C5H9BrMg
Molecular Weight (g/mol) 173.34
MDL Number MFCD00209569
InChI Key PZOJUJCALDFFHC-UHFFFAOYSA-M
Synonym cyclopentylmagnesium bromide, bromocyclopentylmagnesium, magnesium, bromocyclopentyl, cyclopentyl magnesium bromide, bromo cyclopentyl magnesium, cyclopentylmagnesium bromide solution, cydopentylmagnesium bromide, cyclopentylmagnsium bromide, pzojujcaldffhc-uhfffaoysa-m, cyclopentylmagnesium bromide solution, 2.0 m in diethyl ether, grignard reagent
PubChem CID 118418
IUPAC Name magnesium;cyclopentane;bromide
SMILES Br[Mg]C1CCCC1

Specifications

Color Gray
Density 0.9960g/mL
Boiling Point 35°C
Flash Point -38°C
Packaging AcroSeal™ Glass Bottle
Quantity 100 mL
Specific Gravity 0.996
Formula Weight 173.33
Physical Form Solution
Chemical Name or Material Cyclopentylmagnesium bromide, 2.0M solution in diethyl ether
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. Extremely flammable liquid and vapor. In contact with water releases flammable gas. May form explosive peroxides. Reacts violently with water. Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement
Wear eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Avoid breathing dust/fume/gas/mist/vapors/spray. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Store in a dry place. Store in a closed container.
EINECSNumber 251-420-8

RUO – Research Use Only