accessibility menu, dialog, popup

UserName

MgBr

CAS RN 33240-34-5

IUPAC Name: bromo(cyclopentyl)magnesium
Synonyms: bromo(cyclopentyl)magnesium
Molecular Weight (g/mol): 173.34
Molecular Formula: C5H9BrMg
InChi Key: PZOJUJCALDFFHC-UHFFFAOYSA-M
SMILES: Br[Mg]C1CCCC1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)