missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentylacetylene, 95%

Catalog No. p-3212565
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC381310010 1 g Glass bottle
AC381310050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC381310010 Supplier Thermo Scientific Chemicals Supplier No. 381310010

CAS: 930-51-8 | C7H10 | 94.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 930-51-8
Molecular Formula C7H10
Molecular Weight (g/mol) 94.16
MDL Number MFCD00013744
InChI Key TXVJSWLZYQMWPC-UHFFFAOYSA-N
Synonym cyclopentylacetylene, cyclopentyl acetylene, cyclopentylethyne, cyclopentane, ethynyl, cyclopentane, ethynyl-7ci, 8ci, 9ci, ethynylcylopentane, ethynyl-cyclopentane, cyclopentyl-acetylen
PubChem CID 136725
IUPAC Name ethynylcyclopentane
SMILES C#CC1CCCC1

Specifications

Color Yellow
Boiling Point 107°C to 109°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4470 to 1.449
Linear Formula CH∽CC5H9
Quantity 1 g
Formula Weight 94.16
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Cyclopentylacetylene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only