missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentyl Phenyl Ketone 98.0+%, TCI America™

Catalog No. C08825ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C08825ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C08825ML Supplier TCI America Supplier No. C08825ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5422-88-8
Molecular Formula C12H14O
Molecular Weight (g/mol) 174.24
MDL Number MFCD00001378
InChI Key VYDIMQRLNMMJBW-UHFFFAOYSA-N
Synonym cyclopentyl phenyl ketone, cyclopentyl phenyl methanone, benzoylcyclopentane, methanone, cyclopentylphenyl, ketone, cyclopentyl phenyl, cyclopentyl-phenyl-methanone, cyclopentylphenylketone, acmc-209leg, phenyl cyclopentyl ketone, cyclopentanylphenylmethanone
PubChem CID 79464
IUPAC Name cyclopentyl(phenyl)methanone
SMILES O=C(C1CCCC1)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 159°C
Quantity 5 mL
Formula Weight 174.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclopentyl Phenyl Ketone