accessibility menu, dialog, popup

UserName

CAS RN 5422-88-8

O

CAS RN 5422-88-8

IUPAC Name: cyclopentyl(phenyl)methanone
Synonyms: cyclopentyl(phenyl)methanone
Molecular Weight (g/mol): 174.24
Molecular Formula: C12H14O
InChi Key: VYDIMQRLNMMJBW-UHFFFAOYSA-N
SMILES: O=C(C1CCCC1)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)