missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopentyl phenyl ketone, 96%
CAS: 5422-88-8 | C12H14O | 174.24 g/mol
$60.90 - $161.52
Chemical Identifiers
| CAS | 5422-88-8 |
|---|---|
| Molecular Formula | C12H14O |
| Molecular Weight (g/mol) | 174.24 |
| MDL Number | MFCD00001378 |
| InChI Key | VYDIMQRLNMMJBW-UHFFFAOYSA-N |
| Synonym | cyclopentyl phenyl ketone, cyclopentyl phenyl methanone, benzoylcyclopentane, methanone, cyclopentylphenyl, ketone, cyclopentyl phenyl, cyclopentyl-phenyl-methanone, cyclopentylphenylketone, acmc-209leg, phenyl cyclopentyl ketone, cyclopentanylphenylmethanone |
| PubChem CID | 79464 |
| IUPAC Name | cyclopentyl(phenyl)methanone |
| SMILES | O=C(C1CCCC1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1997603
|
Thermo Scientific Chemicals
A1997603 |
1 g |
Each for $60.90
|
|
|||||
|
AAA1997606
|
Thermo Scientific Chemicals
A1997606 |
5 g |
Each for $161.52
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 5422-88-8 | |
| 174.24 | |
| VYDIMQRLNMMJBW-UHFFFAOYSA-N | |
| 79464 | |
| O=C(C1CCCC1)C1=CC=CC=C1 |
| C12H14O | |
| MFCD00001378 | |
| cyclopentyl phenyl ketone, cyclopentyl phenyl methanone, benzoylcyclopentane, methanone, cyclopentylphenyl, ketone, cyclopentyl phenyl, cyclopentyl-phenyl-methanone, cyclopentylphenylketone, acmc-209leg, phenyl cyclopentyl ketone, cyclopentanylphenylmethanone | |
| cyclopentyl(phenyl)methanone |
Specifications
| 5422-88-8 | |
| 136°C to 140°C (16 mmHg) | |
| C12H14O | |
| MFCD00001378 | |
| 2045903 | |
| VYDIMQRLNMMJBW-UHFFFAOYSA-N | |
| cyclopentyl(phenyl)methanone | |
| 79464 | |
| 96% |
| 1.036 | |
| >110°C (230°F) | |
| 1.544 | |
| 1 g | |
| cyclopentyl phenyl ketone, cyclopentyl phenyl methanone, benzoylcyclopentane, methanone, cyclopentylphenyl, ketone, cyclopentyl phenyl, cyclopentyl-phenyl-methanone, cyclopentylphenylketone, acmc-209leg, phenyl cyclopentyl ketone, cyclopentanylphenylmethanone | |
| O=C(C1CCCC1)C1=CC=CC=C1 | |
| 174.24 | |
| 174.24 | |
| Cyclopentyl phenyl ketone |
Safety and Handling
EINECSNumber : 226-548-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only