missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentyl methyl ether, 99.9+%, extra pure, stabilized

Catalog No. AC396130500
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC396130500 50 mL Glass bottle
AC396132500 250 mL Glass bottle
AC396130010 1 L Glass bottle
AC396130025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC396130500 Supplier Thermo Scientific Chemicals Supplier No. 396130500

CAS: 5614-37-9 | C6H12O | 100.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5614-37-9
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD08276401
InChI Key SKTCDJAMAYNROS-UHFFFAOYSA-N
Synonym cyclopentyl methyl ether, cyclopentane, methoxy, cpme, cyclopentane,methoxy, unii-4067e5gbkb, cyclopentylmethylether, cylopentylmethylether, methoxycyclopentane, 2-methoxycyclopentyl, cylopentyl-methylether
PubChem CID 138539
IUPAC Name methoxycyclopentane
SMILES COC1CCCC1

Specifications

Melting Point -140.0°C
Density 0.8600g/mL
Boiling Point 106.0°C
Flash Point −1°C
Infrared Spectrum Authentic
Assay Percent Range 99.9% min. (GC)
Packaging Glass bottle
Refractive Index 1.4190 to 1.4220 (20°C, 589nm)
Linear Formula C5H9OCH3
Quantity 50 mL
Residue after Evaporation 0.01% max.
Merck Index 15,2742
Specific Gravity 0.86
Stabilizer 50ppm (BHT) approximately
Formula Weight 100.16
Percent Purity 99.9+%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Cyclopentyl methyl ether, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May form explosive peroxides.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye protection/face p

RUO – Research Use Only