Learn More
Cyclopentyl methyl ether, 99.5%, Extra Dry, stabilized, AcroSeal™
CAS: 5614-37-9 | C6H12O | 100.16 g/mol
Supplier: Thermo Scientific Chemicals 397250010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Cyclopentyl methyl ether | |
| 5614-37-9 | |
| 0.8600g/mL | |
| −1°C | |
| AcroSeal™ Glass Bottle | |
| C5H9OCH3 | |
| 1 L | |
| 0.005% max. | |
| 15, 2742 | |
| SKTCDJAMAYNROS-UHFFFAOYSA-N | |
| methoxycyclopentane | |
| 50ppm (BHT) approximately | |
| 100.16 | |
| Extra Dry |
| Stabilized, Extra Dry | |
| -140.0°C | |
| 106.0°C | |
| 99.4% min. (GC) | |
| C6H12O | |
| MFCD08276401 | |
| 1993 | |
| 0.86 | |
| cyclopentyl methyl ether, cyclopentane, methoxy, cpme, cyclopentane,methoxy, unii-4067e5gbkb, cyclopentylmethylether, cylopentylmethylether, methoxycyclopentane, 2-methoxycyclopentyl, cylopentyl-methylether | |
| COC1CCCC1 | |
| 100.16 | |
| 138539 | |
| 99.5% | |
| Liquid |
Chemical Identifiers
| 5614-37-9 | |
| 100.16 | |
| SKTCDJAMAYNROS-UHFFFAOYSA-N | |
| 138539 | |
| COC1CCCC1 |
| C6H12O | |
| MFCD08276401 | |
| cyclopentyl methyl ether, cyclopentane, methoxy, cpme, cyclopentane,methoxy, unii-4067e5gbkb, cyclopentylmethylether, cylopentylmethylether, methoxycyclopentane, 2-methoxycyclopentyl, cylopentyl-methylether | |
| methoxycyclopentane |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May form explosive peroxides.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye protection/face p
GHS Signal Word: Danger
RUO – Research Use Only