missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanone, 99+%, pure

CAS: 120-92-3 | C5H8O | 84.12 g/mol

Supplier:  Thermo Scientific Chemicals 111530010

 View more versions of this product

Catalog No. AC111530010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cyclopentanone
-51.0°C
0.9500g/mL
26°C
99% min. (GC)
C5H8O
C5H8(=O)
1 L
0.95
adipic ketone, ketocyclopentane, ketopentamethylene, adipinketon, dumasin, cyclopentanon, oxocyclopentane, cyclopentan-1-one, unii-220w81tn3s, cyclopentyloxy
BGTOWKSIORTVQH-UHFFFAOYSA-N
cyclopentanone
8452
CHEBI:16486
99+%
Liquid
120-92-3
Colorless to Yellow
130.0°C to 131.0°C
Authentic
Glass bottle
1.4360 to 1.4380
MFCD00001409
07,5
15,2738
Solubility in water: 301g/L (20°C). Other solubilities: miscible with alcohol,acetone and ether
O=C1CCCC1
84.12
0.8 mPa.s (20°C)
84.12
Pure

Chemical Identifiers

120-92-3
84.12
BGTOWKSIORTVQH-UHFFFAOYSA-N
8452
cyclopentanone
C5H8O
MFCD00001409
adipic ketone, ketocyclopentane, ketopentamethylene, adipinketon, dumasin, cyclopentanon, oxocyclopentane, cyclopentan-1-one, unii-220w81tn3s, cyclopentyloxy
CHEBI:16486
O=C1CCCC1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 204-435-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only