missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanone, 99+%, pure

Catalog No. p-4258916
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC111532500 250 mL Glass bottle
AC111530010 1 L Glass bottle
AC111530025 2.5 L Glass bottle
AC111530100 10 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC111532500 Supplier Thermo Scientific Chemicals Supplier No. 111532500
Only null left

CAS: 120-92-3 | C5H8O | 84.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 120-92-3
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00001409
InChI Key BGTOWKSIORTVQH-UHFFFAOYSA-N
Synonym adipic ketone, ketocyclopentane, ketopentamethylene, adipinketon, dumasin, cyclopentanon, oxocyclopentane, cyclopentan-1-one, unii-220w81tn3s, cyclopentyloxy
PubChem CID 8452
ChEBI CHEBI:16486
IUPAC Name cyclopentanone
SMILES O=C1CCCC1

Specifications

Melting Point -51.0°C
Color Colorless to Yellow
Density 0.9500g/mL
Boiling Point 130.0°C to 131.0°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C5H8(=O)
Refractive Index 1.4360 to 1.4380
Quantity 250 mL
Beilstein 07,5
Merck Index 15,2738
Specific Gravity 0.95
Solubility Information Solubility in water: 301g/L (20°C). Other solubilities: miscible with alcohol,acetone and ether
Viscosity 0.8 mPa.s (20°C)
Formula Weight 84.12
Percent Purity 99+%
Grade Pure
Physical Form Liquid
Chemical Name or Material Cyclopentanone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-435-9

RUO – Research Use Only