missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanol 99.0+%, TCI America™

Catalog No. C0509500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0509500ML 500 mL
C050925ML 25 mL
C0509100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0509500ML Supplier TCI America Supplier No. C0509500ML

Chemical Identifiers

CAS 96-41-3
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
MDL Number MFCD00001363
InChI Key XCIXKGXIYUWCLL-UHFFFAOYSA-N
Synonym hydroxycyclopentane, cyclopentyl alcohol, 1-cyclopentanol, unii-1l43q07tbu, dsstox_cid_13371, dsstox_rid_79066, dsstox_gsid_33371, cydopentanol, 1-hydroxycyclopentyl, 1-cyclopentyl alcohol
PubChem CID 7298
ChEBI CHEBI:16133
IUPAC Name cyclopentanol
SMILES C1CCC(C1)O

Specifications

Melting Point -19°C
Color Colorless
Boiling Point 140°C
Quantity 500 mL
UN Number 2244
Formula Weight 86.13
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclopentanol
EINECSNumber (3)-2306
TSCA Yes