accessibility menu, dialog, popup

UserName

OH

CAS RN 96-41-3

IUPAC Name: cyclopentanol
Synonyms: cyclopentanol
Molecular Weight (g/mol): 86.13
Molecular Formula: C5H10O
InChi Key: XCIXKGXIYUWCLL-UHFFFAOYSA-N
SMILES: OC1CCCC1
Boiling Point 
  • (2)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (2)
  • (6)
  • (3)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)