missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanol, 99%

Catalog No. AC111521000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC111521000 100 g
AC111525000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC111521000 Supplier Thermo Scientific Chemicals Supplier No. 111521000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 96-41-3 | C5H10O | 86.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 96-41-3
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00001363
InChI Key XCIXKGXIYUWCLL-UHFFFAOYSA-N
Synonym hydroxycyclopentane, cyclopentyl alcohol, 1-cyclopentanol, unii-1l43q07tbu, dsstox_cid_13371, dsstox_rid_79066, dsstox_gsid_33371, cydopentanol, 1-hydroxycyclopentyl, 1-cyclopentyl alcohol
PubChem CID 7298
ChEBI CHEBI:16133
IUPAC Name cyclopentanol
SMILES C1CCC(C1)O

Specifications

Melting Point -19°C
Color Colorless to Yellow
Density 0.9400g/mL
Boiling Point 139°C to 140°C
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4520 to 1.454
Linear Formula C5H9OH
Quantity 100 g
Beilstein 06, 5
Specific Gravity 0.94
Merck Index 15, 2737
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol
Formula Weight 86.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Cyclopentanol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-504-8
TSCA TSCA

RUO – Research Use Only