missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanethiol 98.0+%, TCI America™

Catalog No. p-7124621
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C08105ML 5 mL
C081025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C08105ML Supplier TCI America Supplier No. C08105ML
Only null left

Chemical Identifiers

CAS 1679-07-8
Molecular Formula C5H10S
Molecular Weight (g/mol) 102.20
MDL Number MFCD00001369
InChI Key WVDYBOADDMMFIY-UHFFFAOYSA-N
Synonym cyclopentyl mercaptan, mercaptocyclopentane, cyclopentylthiol, fema no. 3262, cyclopentyl thiol, cyclopen-tanethiol, cyclopentane thiol, cyclopentylmercaptan, acmc-209dxb, cyclopentanethiol
PubChem CID 15510
IUPAC Name cyclopentanethiol
SMILES SC1CCCC1

Specifications

Color Colorless
Boiling Point 132°C
Quantity 5 mL
UN Number 1993
Formula Weight 102.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclopentanethiol
RTECSNumber GY4701000
TSCA Yes