accessibility menu, dialog, popup

UserName

CAS RN 1679-07-8

IUPAC Name: cyclopentanethiol
Synonyms: cyclopentanethiol
Molecular Weight (g/mol): 102.2
Molecular Formula: C5H10S
InChi Key: WVDYBOADDMMFIY-UHFFFAOYSA-N
SMILES: SC1CCCC1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)