Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentanethiol, 97%

Catalog No. AAL0357714
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0357714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0357714 Supplier Thermo Scientific Chemicals Supplier No. L0357714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 1679-07-8 | C5H10S | 102.20 g/mol

Cyclopentanethiol has used as a food spice. It is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentanethiol has used as a food spice. It is a pharmaceutical intermediate.

Solubility
Slightly soluble in water; soluble in alcohol and oils.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.

Chemical Identifiers

CAS 1679-07-8
Molecular Formula C5H10S
Molecular Weight (g/mol) 102.20
MDL Number MFCD00001369
InChI Key WVDYBOADDMMFIY-UHFFFAOYSA-N
Synonym cyclopentyl mercaptan, mercaptocyclopentane, cyclopentylthiol, fema no. 3262, cyclopentyl thiol, cyclopen-tanethiol, cyclopentane thiol, cyclopentylmercaptan, acmc-209dxb, cyclopentanethiol
PubChem CID 15510
IUPAC Name cyclopentanethiol
SMILES SC1CCCC1

Specifications

Density 0.957
Boiling Point 131°C to 132°C
Flash Point 25°C (77°F)
Odor Stench
Refractive Index 1.49
Quantity 25 g
UN Number UN3336
Beilstein 2343861
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water; soluble in alcohol and oils.
Formula Weight 102.2
Percent Purity 97%
Chemical Name or Material Cyclopentanethiol
Show More Show Less
Hazard Category H226-H302-H315-H319-H335
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: MERCAPTANS, LIQUID, FLAMMABLE, N.O.S.
EINECSNumber 216-841-3
RTECSNumber GY4701000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only