missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanemethanol, 98%, Thermo Scientific™

Catalog No. p-97359
Change view
Click to view available options
Quantity:
10g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC147150100 10g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC147150100 Supplier Thermo Scientific Chemicals Supplier No. 147150100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3637-61-4
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00001384
InChI Key ISQVBYGGNVVVHB-UHFFFAOYSA-N
Synonym cyclopentanemethanol, cyclopentyl-methanol, cyclopentylcarbinol, cyclopentylmethyl alcohol, cyclopentanmethanol, cylcopentanemethanol, cyclopentan-methanol, cyclopentyl methanol, cyclopentyl carbinol, cyclo-pentanemethanol
PubChem CID 77195
IUPAC Name cyclopentylmethanol
SMILES C1CCC(C1)CO

Specifications

Density 0.9200g/mL
Boiling Point 162°C to 163°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4565 to 1.4585
Linear Formula C5H9CH2OH
Quantity 10g
Beilstein 06, 9
Specific Gravity 0.92
Formula Weight 100.16
Percent Purity 98%
Chemical Name or Material Cyclopentanemethanol, 96%
Show More Show Less
EINECSNumber 222-861-3

RUO – Research Use Only