missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanecarboxylic Acid 98.0+%, TCI America™

Catalog No. c05125g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
C05125G 5 g
C051225G 25 g
2 options

Catalog No. C05125G

Supplier: TCI America C05125G

Only null left

Chemical Identifiers

CAS 3400-45-1
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00001371
InChI Key JBDSSBMEKXHSJF-UHFFFAOYSA-N
Synonym cyclopentane carboxylic acid, cyclopentylcarboxylic acid, cyclopentancarboxylic acid, cyclopentanoic acid, unii-z6691vh94a, cyclopentylmethanoic acid, pubchem14153, carboxycyclopentane, cyclopentyl carboxylic acid, acmc-209i4g
PubChem CID 18840
IUPAC Name cyclopentanecarboxylic acid
SMILES C1CCC(C1)C(=O)O

Specifications

Color Yellow
Boiling Point 216°C
Quantity 5 g
Formula Weight 114.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Cyclopentanecarboxylic Acid
EINECSNumber (3)-3821
TSCA No