missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanecarboxylic acid, 99%

CAS: 3400-45-1 | C6H10O2 | 114.144 g/mol

Supplier:  Thermo Scientific Chemicals A1237518

 View more versions of this product

Catalog No. AAA1237518


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cyclopentanecarboxylic acid
-7°C
215°C to 216°C
C6H10O2
MFCD00001371
2039729
JBDSSBMEKXHSJF-UHFFFAOYSA-N
cyclopentanecarboxylic acid
18840
99%
3400-45-1
1.052
93°C (199°F)
1.454
50 g
cyclopentane carboxylic acid, cyclopentylcarboxylic acid, cyclopentancarboxylic acid, cyclopentanoic acid, unii-z6691vh94a, cyclopentylmethanoic acid, pubchem14153, carboxycyclopentane, cyclopentyl carboxylic acid, acmc-209i4g
C1CCC(C1)C(=O)O
114.144
114.14

Chemical Identifiers

3400-45-1
114.144
JBDSSBMEKXHSJF-UHFFFAOYSA-N
18840
C1CCC(C1)C(=O)O
C6H10O2
MFCD00001371
cyclopentane carboxylic acid, cyclopentylcarboxylic acid, cyclopentancarboxylic acid, cyclopentanoic acid, unii-z6691vh94a, cyclopentylmethanoic acid, pubchem14153, carboxycyclopentane, cyclopentyl carboxylic acid, acmc-209i4g
cyclopentanecarboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 222-269-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only