Learn More
Cyclopentanecarboxaldehyde, 98%, stab.
CAS: 872-53-7 | C6H10O | 98.145 g/mol
Supplier: Thermo Scientific Chemicals H5660906
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Soluble in water and ethanol
Notes
Light sensitive. Incompatible with oxidizing agents. Store at room temperature.
Specifications
| Cyclopentanecarboxaldehyde | |
| 872-53-7 | |
| 140°C to 141°C | |
| C6H10O | |
| MFCD00060798 | |
| UN1989 | |
| Soluble in water and ethanol | |
| C1CCC(C1)C=O | |
| 98.145 | |
| 98.14 |
| stabilized with 25ppm 1,4-benzoquinone | |
| 0.919 | |
| 28°C (82°F) | |
| 1.443 | |
| 5 g | |
| cyclopentanecarboxaldehyde, cyclopentanealdehyde, cyclopentancarbaldehyd, cyclopentylformaldehyde, formylcyclopentane, cyclopentyl aldehyde, 1-formylcyclopentane, cyclopentane-1-carboxaldehyde, cyclopentanecaboxaldehyde, cyclopentanecarboaldehyde | |
| VELDYOPRLMJFIK-UHFFFAOYSA-N | |
| cyclopentanecarbaldehyde | |
| 70106 | |
| 98% |
Chemical Identifiers
| 872-53-7 | |
| 98.145 | |
| VELDYOPRLMJFIK-UHFFFAOYSA-N | |
| 70106 | |
| C1CCC(C1)C=O |
| C6H10O | |
| MFCD00060798 | |
| cyclopentanecarboxaldehyde, cyclopentanealdehyde, cyclopentancarbaldehyd, cyclopentylformaldehyde, formylcyclopentane, cyclopentyl aldehyde, 1-formylcyclopentane, cyclopentane-1-carboxaldehyde, cyclopentanecaboxaldehyde, cyclopentanecarboaldehyde | |
| cyclopentanecarbaldehyde |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, N.O.S.
EINECSNumber : 212-829-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only