missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanecarboxaldehyde, 98%, stab.

Catalog No. p-7037794
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5660903 1 g
AAH5660906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. AAH5660903 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' H5660903
Only null left

CAS: 872-53-7 | C6H10O | 98.145 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in water and ethanol

Notes
Light sensitive. Incompatible with oxidizing agents. Store at room temperature.

Chemical Identifiers

CAS 872-53-7
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00060798
InChI Key VELDYOPRLMJFIK-UHFFFAOYSA-N
Synonym cyclopentanecarboxaldehyde, cyclopentanealdehyde, cyclopentancarbaldehyd, cyclopentylformaldehyde, formylcyclopentane, cyclopentyl aldehyde, 1-formylcyclopentane, cyclopentane-1-carboxaldehyde, cyclopentanecaboxaldehyde, cyclopentanecarboaldehyde
PubChem CID 70106
IUPAC Name cyclopentanecarbaldehyde
SMILES C1CCC(C1)C=O

Specifications

Density 0.919
Boiling Point 140°C to 141°C
Flash Point 28°C (82°F)
Refractive Index 1.443
Quantity 1 g
UN Number UN1989
Solubility Information Soluble in water and ethanol
Formula Weight 98.14
Percent Purity 98%
Chemical Name or Material Cyclopentanecarboxaldehyde, stabilized with 25ppm 1,4-benzoquinone
missing translation for 'healthHazard1' H226-H315-H319-H335
missing translation for 'healthHazard2' GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, N.O.S.
EINECSNumber 212-829-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only