missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanecarbonitrile, 98%

Catalog No. AAA1771003
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1771003 1 g
AAA1771006 5 g
AAA1771014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1771003 Supplier Thermo Scientific Chemicals Supplier No. A1771003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4254-02-8 | C6H9N | 95.15 g/mol

Cyclopentanecarbonitrile is used as a substrate and reagent to investigate the specific activity of nitrile and amide hydrolyzing enzymes isolated from candida guilliermondii cells. It is also used to prepare cyclopentanecarboxamide by using tetrabutylammonium hydrogen sulfate as a catalyst in the presence of reagents such as aqueous hydrogen peroxide and sodium hydroxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentanecarbonitrile is used as a substrate and reagent to investigate the specific activity of nitrile and amide hydrolyzing enzymes isolated from candida guilliermondii cells. It is also used to prepare cyclopentanecarboxamide by using tetrabutylammonium hydrogen sulfate as a catalyst in the presence of reagents such as aqueous hydrogen peroxide and sodium hydroxide.

Solubility
Miscible with toluene.

Notes
Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 4254-02-8
MDL Number MFCD00013741

Specifications

Melting Point -76°C
Density 0.912
Boiling Point 67°C to 68°C (10 mmHg)
Flash Point 56°C (133°F)
Molecular Formula C2H6O6Zn
Refractive Index 1.441
Quantity 1 g
UN Number UN1992
Beilstein 1098717
Synonym zinc oxalate dihydrate, zinc oxalate, c2o4zn.2h2o
Solubility Information Miscible with toluene.
InChI Key PLYRAVNBPDIEOH-UHFFFAOYSA-N
SMILES C(=O)(C(=O)O)O.O.O.[Zn]
IUPAC Name oxalic acid;zinc;dihydrate
Molecular Weight (g/mol) 191.44
PubChem CID 91886451
Formula Weight 95.14
Percent Purity 98%
Chemical Name or Material Cyclopentanecarbonitrile
Show More Show Less
Hazard Category H226-H301-H315-H319-H335
Hazard Statement GHS H Statement
H331-H226-H302-H312-H315-H319-H335
Toxic if inhaled.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 224-229-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only