missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentadienylvanadium tetracarbonyl, 97+%

Catalog No. p-7080351
Change view
Click to view available options
Quantity:
0.5 g
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8112302 0.5 g
AA8112304 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA8112302 Supplier Thermo Scientific Chemicals Supplier No. 08112302
Only null left

CAS: 12108-04-2 | C9H5O4V | 228.076 g/mol

Employed as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12108-04-2
Molecular Formula C9H5O4V
Molecular Weight (g/mol) 228.076
MDL Number MFCD00014259
InChI Key LIFGJKDEFQAOFY-UHFFFAOYSA-N
Synonym carbon monoxide; cyclopentane; vanadium, tetracarbonyl .eta.5-cyclopenta-2,4-dien-1-yl vanadium
PubChem CID 498652
IUPAC Name carbon monoxide;cyclopentane;vanadium
SMILES [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[CH]1[CH][CH][CH][CH]1.[V]

Specifications

Melting Point 138°C
Boiling Point 80°C to 100°C (0.1 mmHg)(sublimation)
Quantity 0.5 g
UN Number UN3466
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 228.08
Percent Purity ≥97%
Physical Form Crystalline
Chemical Name or Material Cyclopentadienylvanadium tetracarbonyl
Hazard Category H301+H311-H315-H319-H330-H335
Hazard Statement GHS H Statement
H300-H310-H330-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
EINECSNumber 235-163-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only