accessibility menu, dialog, popup

UserName

COCOCOCOHCCHCHHCHCV

CAS RN 12108-04-2

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 228.08
Molecular Formula: C9H5O4V
InChi Key: LIFGJKDEFQAOFY-UHFFFAOYSA-N
SMILES: [V].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[CH]1[CH][CH][CH][CH]1
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)