missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentadecane 98.0+%, TCI America™

Catalog No. C12245G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C12245G Supplier TCI America Supplier No. C12245G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 295-48-7
Molecular Formula C15H30
Molecular Weight (g/mol) 210.405
MDL Number MFCD00039424
InChI Key SRONXYPFSAKOGH-UHFFFAOYSA-N
Synonym acmc-209h9p
PubChem CID 67525
IUPAC Name cyclopentadecane
SMILES C1CCCCCCCCCCCCCC1

Specifications

Melting Point 64°C
Color White
Quantity 5 g
Formula Weight 210.41
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Cyclopentadecane