accessibility menu, dialog, popup

UserName

CAS RN 295-48-7

IUPAC Name: cyclopentadecane
Synonyms: cyclopentadecane
Molecular Weight (g/mol): 210.41
Molecular Formula: C15H30
InChi Key: SRONXYPFSAKOGH-UHFFFAOYSA-N
SMILES: C1CCCCCCCCCCCCCC1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)