missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentadecane, 98%

Catalog No. p-7024554
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1941603 1 g
AAA1941606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1941603 Supplier Thermo Scientific Chemicals Supplier No. A1941603
Only null left

CAS: 295-48-7 | C15H30 | 210.405 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 295-48-7
Molecular Formula C15H30
Molecular Weight (g/mol) 210.405
MDL Number MFCD00039424
InChI Key SRONXYPFSAKOGH-UHFFFAOYSA-N
Synonym acmc-209h9p
PubChem CID 67525
IUPAC Name cyclopentadecane
SMILES C1CCCCCCCCCCCCCC1

Specifications

Density 0.805
Quantity 1 g
Formula Weight 210.4
Percent Purity 98%
Chemical Name or Material Cyclopentadecane
EINECSNumber 206-040-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only