missing translation for 'onlineSavingsMsg'
Learn More

Cyclopamine, 99%

CAS: 4449-51-8 | C27H41NO2 | 411.63 g/mol

$568.42 - $963.86

Chemical Identifiers

CAS 4449-51-8
Molecular Formula C27H41NO2
Molecular Weight (g/mol) 411.63
MDL Number MFCD01735266
InChI Key QASFUMOKHFSJGL-LAFRSMQTSA-N
Synonym cyclopamine, 3b,23b-17,23-epoxy-11-deoxoveratraman-3-ol
PubChem CID 122130428
SMILES C[C@@H]1[C@@H]2NC[C@@H](C)C[C@H]2O[C@]11CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC2=C1C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61528MB
View Documents
Thermo Scientific Chemicals
J61528MB
25 mg
Each for $568.42
Only null left
 
AAJ61528MF
View Documents
Thermo Scientific Chemicals
J61528MF
50 mg
Each for $963.86
Only null left
 
Description

Description

Cyclopamine, is used as a hedgehog [Hh] signaling pathway and Smo inhibitor. It depletes stem-like cancer cells in glioblastoma and blocks tumor engraftment. Can also be used to induce differentiation of human embryonic stem cells (hESCs) into hormone-expressing endocrine cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopamine, is used as a hedgehog [Hh] signaling pathway and Smo inhibitor. It depletes stem-like cancer cells in glioblastoma and blocks tumor engraftment. Can also be used to induce differentiation of human embryonic stem cells (hESCs) into hormone-expressing endocrine cells.

Solubility
Soluble in ethanol (5 mg/ml warm), DMSO (4 mg/ml), DMF (10 mg/ml), methylene chloride, chloroform, methanol (7 mg/ml), and benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

4449-51-8
411.63
QASFUMOKHFSJGL-LAFRSMQTSA-N
122130428
C27H41NO2
MFCD01735266
cyclopamine, 3b,23b-17,23-epoxy-11-deoxoveratraman-3-ol
C[C@@H]1[C@@H]2NC[C@@H](C)C[C@H]2O[C@]11CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC2=C1C

Specifications

4449-51-8
White
MFCD01735266
cyclopamine, 3b,23b-17,23-epoxy-11-deoxoveratraman-3-ol
QASFUMOKHFSJGL-LAFRSMQTSA-N
(3S,3'R,3'aS,6'S,6aS,9S)-3',6',10,11b-tetramethylspiro[2,3,4,6,6a,6b,7,8,11,11a-decahydro-1H-benzo[a]fluorene-9,2'-3a,4,5,6,7,7a-hexahydro-3H-furo[3,2-b]pyridine]-3-ol
122130428
≥99%
Cyclopamine
234°C to 237°C
C27H41NO2
25 mg
Soluble in ethanol (5mg/ml warm),DMSO (4mg/ml),DMF (10mg/ml),methylene chloride,chloroform,methanol (7mg/ml),and benzene.
C[C@@H]1[C@@H]2NC[C@@H](C)C[C@H]2O[C@]11CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC2=C1C
411.63
411.62
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H341-H361
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.

P201-P202-P281-P308+P313-P501c

H341-H361

RTECSNumber : GY0750000

TSCA : No

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only