missing translation for 'onlineSavingsMsg'
Learn More

Cyclopamine, 99%

Catalog No. AAJ61528MB
Change view
Click to view available options
Quantity:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61528MB 25 mg
AAJ61528MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61528MB Supplier Thermo Scientific Chemicals Supplier No. J61528MB
Only null left

CAS: 4449-51-8 | C27H41NO2 | 411.63 g/mol

Cyclopamine, is used as a hedgehog [Hh] signaling pathway and Smo inhibitor. It depletes stem-like cancer cells in glioblastoma and blocks tumor engraftment. Can also be used to induce differentiation of human embryonic stem cells (hESCs) into hormone-expressing endocrine cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopamine, is used as a hedgehog [Hh] signaling pathway and Smo inhibitor. It depletes stem-like cancer cells in glioblastoma and blocks tumor engraftment. Can also be used to induce differentiation of human embryonic stem cells (hESCs) into hormone-expressing endocrine cells.

Solubility
Soluble in ethanol (5 mg/ml warm), DMSO (4 mg/ml), DMF (10 mg/ml), methylene chloride, chloroform, methanol (7 mg/ml), and benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 4449-51-8
Molecular Formula C27H41NO2
Molecular Weight (g/mol) 411.63
MDL Number MFCD01735266
InChI Key QASFUMOKHFSJGL-LAFRSMQTSA-N
Synonym cyclopamine, 3b,23b-17,23-epoxy-11-deoxoveratraman-3-ol
PubChem CID 122130428
SMILES C[C@@H]1[C@@H]2NC[C@@H](C)C[C@H]2O[C@]11CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC2=C1C

Specifications

Melting Point 234°C to 237°C
Color White
Quantity 25 mg
Solubility Information Soluble in ethanol (5mg/ml warm),DMSO (4mg/ml),DMF (10mg/ml),methylene chloride,chloroform,methanol (7mg/ml),and benzene.
IUPAC Name (3S,3'R,3'aS,6'S,6aS,9S)-3',6',10,11b-tetramethylspiro[2,3,4,6,6a,6b,7,8,11,11a-decahydro-1H-benzo[a]fluorene-9,2'-3a,4,5,6,7,7a-hexahydro-3H-furo[3,2-b]pyridine]-3-ol
Formula Weight 411.62
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material Cyclopamine
Hazard Category H341-H361
Hazard Statement GHS H Statement
H341-H361
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
Precautionary Statement P201-P202-P281-P308+P313-P501c
RTECSNumber GY0750000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only