missing translation for 'onlineSavingsMsg'
Learn More

Cyclooctene, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020165
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020165 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020165 Supplier Merck Emd Millipore Supplier No. 296481ML
Only null left
Cleaning Products, Cosmetics, Environmental, Food and Beverages, Personal Care and Petroleum

Specifications

CAS 931-88-4
Density 0.848 g/mL (at 20°C)
Flash Point 24°C - Closed Cup
Physical Form Neat
Linear Formula C8H14
MDL Number MFCD00001753
Boiling Point 32°C to 34°C (12 mmHg)
Type Analytical Standard
Quantity 1 mL
Formula Weight 110.20
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less