missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclooctanone oxime, 98+%
CAS: 1074-51-7 | C8H15NO | 141.21 g/mol
Supplier: Thermo Scientific Chemicals A1667218
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Cyclooctanone oxime | |
| 36°C to 37°C | |
| C8H15NO | |
| 50 g | |
| KTPUHSVFNHULJH-UHFFFAOYSA-N | |
| N-cyclooctylidenehydroxylamine | |
| 136841 | |
| ≥98% |
| 1074-51-7 | |
| 128°C to 129°C (14 mmHg) | |
| MFCD00191967 | |
| cyclooctanone oxime, cyclooctanone, oxime, cyclooctanonoxim, cyclooctanoneoxime, acmc-1bxvz, hydroxyimino cyclooctane | |
| ON=C1CCCCCCC1 | |
| 141.21 | |
| 141.22 |
Chemical Identifiers
| 1074-51-7 | |
| 141.21 | |
| KTPUHSVFNHULJH-UHFFFAOYSA-N | |
| 136841 | |
| ON=C1CCCCCCC1 |
| C8H15NO | |
| MFCD00191967 | |
| cyclooctanone oxime, cyclooctanone, oxime, cyclooctanonoxim, cyclooctanoneoxime, acmc-1bxvz, hydroxyimino cyclooctane | |
| N-cyclooctylidenehydroxylamine |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only