missing translation for 'onlineSavingsMsg'
Learn More

Cyclooctanone oxime, 98+%

Catalog No. p-7023279
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1667209 10 g
AAA1667218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1667209 Supplier Thermo Scientific Chemicals Supplier No. A1667209
Only null left

CAS: 1074-51-7 | C8H15NO | 141.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1074-51-7
Molecular Formula C8H15NO
Molecular Weight (g/mol) 141.21
MDL Number MFCD00191967
InChI Key KTPUHSVFNHULJH-UHFFFAOYSA-N
Synonym cyclooctanone oxime, cyclooctanone, oxime, cyclooctanonoxim, cyclooctanoneoxime, acmc-1bxvz, hydroxyimino cyclooctane
PubChem CID 136841
IUPAC Name N-cyclooctylidenehydroxylamine
SMILES ON=C1CCCCCCC1

Specifications

Melting Point 36°C to 37°C
Boiling Point 128°C to 129°C (14 mmHg)
Quantity 10 g
Formula Weight 141.22
Percent Purity ≥98%
Chemical Name or Material Cyclooctanone oxime
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only