missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexylurea, 98%
CAS: 698-90-8 | C7H14N2O | 142.202 g/mol
$58.42 - $165.22
Chemical Identifiers
| CAS | 698-90-8 |
|---|---|
| Molecular Formula | C7H14N2O |
| Molecular Weight (g/mol) | 142.202 |
| MDL Number | MFCD00014286 |
| InChI Key | WUESWDIHTKHGQA-UHFFFAOYSA-N |
| Synonym | 1-cyclohexylurea, urea, cyclohexyl, n-cyclohexylurea, n-cyclohexlurea, cyclohexyl urea, amino-n-cyclohexylamide, cyclohexyl-urea, monocyclohexylurea, n'-cyclohexylurea, n'-cyclohexyl urea |
| PubChem CID | 69684 |
| IUPAC Name | cyclohexylurea |
| SMILES | C1CCC(CC1)NC(=O)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2491514
|
Thermo Scientific Chemicals
B2491514 |
25 g |
Each for $58.42
|
|
|||||
|
AAB2491522
|
Thermo Scientific Chemicals
B2491522 |
100 g |
Each for $165.22
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 698-90-8 | |
| 142.202 | |
| WUESWDIHTKHGQA-UHFFFAOYSA-N | |
| 69684 | |
| C1CCC(CC1)NC(=O)N |
| C7H14N2O | |
| MFCD00014286 | |
| 1-cyclohexylurea, urea, cyclohexyl, n-cyclohexylurea, n-cyclohexlurea, cyclohexyl urea, amino-n-cyclohexylamide, cyclohexyl-urea, monocyclohexylurea, n'-cyclohexylurea, n'-cyclohexyl urea | |
| cyclohexylurea |
Specifications
| 698-90-8 | |
| C7H14N2O | |
| 25 g | |
| 1-cyclohexylurea, urea, cyclohexyl, n-cyclohexylurea, n-cyclohexlurea, cyclohexyl urea, amino-n-cyclohexylamide, cyclohexyl-urea, monocyclohexylurea, n'-cyclohexylurea, n'-cyclohexyl urea | |
| C1CCC(CC1)NC(=O)N | |
| 142.202 | |
| 142.2 | |
| Cyclohexylurea |
| 191°C to 195°C | |
| MFCD00014286 | |
| 636914 | |
| WUESWDIHTKHGQA-UHFFFAOYSA-N | |
| cyclohexylurea | |
| 69684 | |
| 98% |
Safety and Handling
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 211-822-6
RTECSNumber : YS7564300
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only