accessibility menu, dialog, popup

UserName

NH2ONH

CAS RN 698-90-8

IUPAC Name: cyclohexylurea
Synonyms: urea, cyclohexyl-
Molecular Weight (g/mol): 142.2
Molecular Formula: C7H14N2O
InChi Key: WUESWDIHTKHGQA-UHFFFAOYSA-N
SMILES: NC(=O)NC1CCCCC1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)