missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyltrimethoxysilane 97.0+%, TCI America™

Catalog No. C22805G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22805G 5 g
C228025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22805G Supplier TCI America Supplier No. C22805G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17865-54-2
Molecular Formula C9H20O3Si
Molecular Weight (g/mol) 204.34
MDL Number MFCD01074466
InChI Key MEWFSXFFGFDHGV-UHFFFAOYSA-N
PubChem CID 10998130
IUPAC Name cyclohexyltrimethoxysilane
SMILES CO[Si](OC)(OC)C1CCCCC1

Specifications

Color Yellow
Boiling Point 208°C
Quantity 5 g
Formula Weight 204.34
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclohexyltrimethoxysilane