accessibility menu, dialog, popup

UserName

CAS RN 17865-54-2

IUPAC Name: cyclohexyltrimethoxysilane
Synonyms: cyclohexyltrimethoxysilane
Molecular Weight (g/mol): 204.34
Molecular Formula: C9H20O3Si
InChi Key: MEWFSXFFGFDHGV-UHFFFAOYSA-N
SMILES: CO[Si](OC)(OC)C1CCCCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results