missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylmethyl isothiocyanate, 97%

Catalog No. AAL1012903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1012903 1 g
AAL1012906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1012903 Supplier Thermo Scientific Chemicals Supplier No. L1012903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 52395-66-1 | C8H13NS | 155.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52395-66-1
Molecular Formula C8H13NS
Molecular Weight (g/mol) 155.26
MDL Number MFCD00040881
InChI Key RYBJNWIQNLZNPE-UHFFFAOYSA-N
Synonym cyclohexanemethyl isothiocyanate, isothiocyanatomethyl cyclohexane, cyclohexylmethyl isothiocyanate, cyclohexylmethanisothiocyanate, cyclohexanemethylisothiocyanate, isothiocyanatomethyl cyclohexane #
PubChem CID 142928
IUPAC Name isothiocyanatomethylcyclohexane
SMILES S=C=NCC1CCCCC1

Specifications

Density 1.02
Boiling Point 222°C to 223°C
Flash Point >110°C (230°F)
Refractive Index 1.536
Quantity 1 g
UN Number UN2922
Beilstein 2079123
Sensitivity Moisture sensitive
Formula Weight 155.26
Percent Purity 97%
Chemical Name or Material Cyclohexylmethyl isothiocyanate
Show More Show Less
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only