Learn More
Cyclohexylmethyl bromide, 96%
CAS: 2550-36-9 | C7H13Br | 177.09 g/mol
$135.73 - $135.73
Chemical Identifiers
| CAS | 2550-36-9 |
|---|---|
| Molecular Formula | C7H13Br |
| Molecular Weight (g/mol) | 177.09 |
| MDL Number | MFCD00001509 |
| InChI Key | UUWSLBWDFJMSFP-UHFFFAOYSA-N |
| Synonym | bromomethyl cyclohexane, cyclohexylmethyl bromide, cyclohexane, bromomethyl, bromocyclohexylmethane, cyclohexylmethylbromide, cyclohexylmethybromide, bromomethyl-cyclohexane, pubchem3103, 1-bromomethylcyclohexane |
| PubChem CID | 137636 |
| SMILES | BrCC1CCCCC1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC111350250
|
Thermo Scientific Chemicals
111350250 |
25 g | Glass bottle |
Each for $135.73
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2550-36-9 | |
| 177.09 | |
| UUWSLBWDFJMSFP-UHFFFAOYSA-N | |
| 137636 |
| C7H13Br | |
| MFCD00001509 | |
| bromomethyl cyclohexane, cyclohexylmethyl bromide, cyclohexane, bromomethyl, bromocyclohexylmethane, cyclohexylmethylbromide, cyclohexylmethybromide, bromomethyl-cyclohexane, pubchem3103, 1-bromomethylcyclohexane | |
| BrCC1CCCCC1 |
Specifications
| 2550-36-9 | |
| 1.2800g/mL | |
| 57°C | |
| 95% min. (GC) | |
| C7H13Br | |
| C6H11CH2Br | |
| 25 g | |
| 1.28 | |
| UUWSLBWDFJMSFP-UHFFFAOYSA-N | |
| bromomethylcyclohexane | |
| 137636 | |
| 96% | |
| Cyclohexylmethyl bromide |
| Yellow | |
| 76°C to 77°C (26.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4910 to 1.493 | |
| MFCD00001509 | |
| 05, IV, 100 | |
| bromomethyl cyclohexane, cyclohexylmethyl bromide, cyclohexane, bromomethyl, bromocyclohexylmethane, cyclohexylmethylbromide, cyclohexylmethybromide, bromomethyl-cyclohexane, pubchem3103, 1-bromomethylcyclohexane | |
| BrCC1CCCCC1 | |
| 177.09 | |
| 177.08 | |
| Liquid |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only