missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexylmethyl Acetate 98.0+%, TCI America™
$201.32 - $609.54
Chemical Identifiers
| CAS | 937-55-3 |
|---|---|
| Molecular Formula | C9H16O2 |
| Molecular Weight (g/mol) | 156.23 |
| MDL Number | MFCD07780584 |
| InChI Key | FXKHUBNHBYCRNH-UHFFFAOYSA-N |
| Synonym | Acetic Acid Cyclohexylmethyl Ester |
| PubChem CID | 244762 |
| IUPAC Name | cyclohexylmethyl acetate |
| SMILES | CC(=O)OCC1CCCCC1 |
Chemical Identifiers
| 937-55-3 | |
| 156.23 | |
| FXKHUBNHBYCRNH-UHFFFAOYSA-N | |
| 244762 | |
| CC(=O)OCC1CCCCC1 |
| C9H16O2 | |
| MFCD07780584 | |
| Acetic Acid Cyclohexylmethyl Ester | |
| cyclohexylmethyl acetate |
Specifications
| 937-55-3 | |
| 183°C | |
| MFCD07780584 | |
| Acetic Acid Cyclohexylmethyl Ester | |
| CC(=O)OCC1CCCCC1 | |
| 156.23 | |
| 156.23 | |
| Liquid |
| Yellow | |
| C9H16O2 | |
| 5 g | |
| FXKHUBNHBYCRNH-UHFFFAOYSA-N | |
| cyclohexylmethyl acetate | |
| 244762 | |
| ≥98.0% (GC) | |
| Cyclohexylmethyl Acetate |
Safety and Handling
TSCA : No