CAS RN 937-55-3
CAS RN 937-55-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cyclohexylmethyl Acetate 98.0+%, TCI America™
CAS: 937-55-3 Molecular Formula: C9H16O2 Molecular Weight (g/mol): 156.23 MDL Number: MFCD07780584 InChI Key: FXKHUBNHBYCRNH-UHFFFAOYSA-N Synonym: Acetic Acid Cyclohexylmethyl Ester PubChem CID: 244762 IUPAC Name: cyclohexylmethyl acetate SMILES: CC(=O)OCC1CCCCC1