missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylmagnesium chloride, 1.3M solution in THF/toluene, AcroSeal™

CAS: 931-51-1 | C6H11ClMg | 142.91 g/mol

$106.47 - $106.47

Chemical Identifiers

CAS 931-51-1
Molecular Formula C6H11ClMg
Molecular Weight (g/mol) 142.91
MDL Number MFCD00003816
InChI Key WMJMABVHDMRMJA-UHFFFAOYSA-M
Synonym cyclohexylmagnesium chloride, magnesium, chlorocyclohexyl, magnesium,chlorocyclohexyl, cyclohexyl magnesium chloride, chloro cyclohexyl magnesium, chlorocyclohexylmagnesium, cyclohexylmagnesiumchloride, unii-oag686rdsu, cyclohexylmagnesium chloride solution, 2.0 m in diethyl ether, cyclohexylmagnesium cloride
PubChem CID 13605
IUPAC Name magnesium;cyclohexane;chloride
SMILES Cl[Mg]C1CCCCC1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC377341000
SDS View Documents
Thermo Scientific Chemicals
377341000
100 mL AcroSeal™ Glass bottle
Each for $106.47
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

931-51-1
142.91
WMJMABVHDMRMJA-UHFFFAOYSA-M
13605
Cl[Mg]C1CCCCC1
C6H11ClMg
MFCD00003816
cyclohexylmagnesium chloride, magnesium, chlorocyclohexyl, magnesium,chlorocyclohexyl, cyclohexyl magnesium chloride, chloro cyclohexyl magnesium, chlorocyclohexylmagnesium, cyclohexylmagnesiumchloride, unii-oag686rdsu, cyclohexylmagnesium chloride solution, 2.0 m in diethyl ether, cyclohexylmagnesium cloride
magnesium;cyclohexane;chloride

Specifications

931-51-1,109-99-9,108-88-3
0.9500g/mL
AcroSeal™ Glass bottle
(C6H11)MgCl
100 mL
cyclohexylmagnesium chloride, magnesium, chlorocyclohexyl, magnesium,chlorocyclohexyl, cyclohexyl magnesium chloride, chloro cyclohexyl magnesium, chlorocyclohexylmagnesium, cyclohexylmagnesiumchloride, unii-oag686rdsu, cyclohexylmagnesium chloride solution, 2.0 m in diethyl ether, cyclohexylmagnesium cloride
WMJMABVHDMRMJA-UHFFFAOYSA-M
magnesium;cyclohexane;chloride
13605
Solution
Brown
−21°C
C6H11ClMg
MFCD00003816
0.95
Solubility in water: reacts violently with water
Cl[Mg]C1CCCCC1
142.91
142.91
Cyclohexylmagnesium chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Suspected of damag

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

EINECSNumber : 213-237-1

SDS
Documents

Documents

RUO – Research Use Only