missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylboronic acid pinacol ester, 97%

Catalog No. p-7055567
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6417803 1 g
AAH6417806 5 g
AAH6417814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6417803 Supplier Thermo Scientific Chemicals Supplier No. H6417803
Only null left

CAS: 87100-15-0 | C12H23BO2 | 210.124 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 87100-15-0
Molecular Formula C12H23BO2
Molecular Weight (g/mol) 210.124
MDL Number MFCD04038749
InChI Key OUEVCDGYTKLNMJ-UHFFFAOYSA-N
Synonym cyclohexylboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-cyclohexyl-4,4,5,5-tetramethyl, cyclohexylboronic acid, pinacol ester, 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl cyclohexane, pubchem7924, cyclohexylboronic acid, pinacol eser, 1-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl cyclohexane
PubChem CID 3668596
IUPAC Name 2-cyclohexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2CCCCC2

Specifications

Color Colorless
Quantity 1 g
Formula Weight 210.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Cyclohexylboronic acid pinacol ester
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only