accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3C

CAS RN 87100-15-0

IUPAC Name: 2-cyclohexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-cyclohexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 210.12
Molecular Formula: C12H23BO2
InChi Key: OUEVCDGYTKLNMJ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1CCCCC1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)