Learn More
Cyclohexylbenzene, 97+%
Description
Cyclohexylbenzene is used as a high boiling point solvent. It finds application as a penetrant used in plastics, painting, and adhesives. It acts as an additive for lithium-ion battery electrolyte to enhance the battery safety. It is used to prepare intermediates such as phenol and cyclohexanone. Further, it is used in the preparation of liquid crystal display (LCD) material intermediate 4-ethyl cyclohexyl benzoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Cyclohexylbenzene is used as a high boiling point solvent. It finds application as a penetrant used in plastics, painting, and adhesives. It acts as an additive for lithium-ion battery electrolyte to enhance the battery safety. It is used to prepare intermediates such as phenol and cyclohexanone. Further, it is used in the preparation of liquid crystal display (LCD) material intermediate 4-ethyl cyclohexyl benzoic acid.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 5°C to 6°C |
| Density | 0.945 |
| Boiling Point | 239°C to 240°C |
| Flash Point | 98°C (208°F) |
| Odor | Mild |
| Refractive Index | 1.526 |
| Quantity | 100 g |
| UN Number | UN3082 |
| Beilstein | 1906803 |
| Solubility Information | Immiscible with water. |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319 Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S. |
| EINECSNumber | 212-572-0 |
| RTECSNumber | CZ1330000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only