missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylbenzene, 97+%

Catalog No. AAA1632036
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1632036 500 g
AAA1632022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1632036 Supplier Thermo Scientific Chemicals Supplier No. A1632036
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 827-52-1 | C12H16 | 160.26 g/mol

Cyclohexylbenzene is used as a high boiling point solvent. It finds application as a penetrant used in plastics, painting, and adhesives. It acts as an additive for lithium-ion battery electrolyte to enhance the battery safety. It is used to prepare intermediates such as phenol and cyclohexanone. Further, it is used in the preparation of liquid crystal display (LCD) material intermediate 4-ethyl cyclohexyl benzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexylbenzene is used as a high boiling point solvent. It finds application as a penetrant used in plastics, painting, and adhesives. It acts as an additive for lithium-ion battery electrolyte to enhance the battery safety. It is used to prepare intermediates such as phenol and cyclohexanone. Further, it is used in the preparation of liquid crystal display (LCD) material intermediate 4-ethyl cyclohexyl benzoic acid.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 827-52-1
Molecular Formula C12H16
Molecular Weight (g/mol) 160.26
MDL Number MFCD00001451
InChI Key IGARGHRYKHJQSM-UHFFFAOYSA-N
Synonym phenylcyclohexane, benzene, cyclohexyl, cyclohexane, phenyl, cyclohexyl-benzene, cyclohexyl benzene, 4-cyclohexylbenzene, santosol 360, phenyl cyclohexane, 1,1'-biphenyl, 1,2,3,4,5,6-hexahydro, wln: l6tj ar
PubChem CID 13229
IUPAC Name cyclohexylbenzene
SMILES C1CCC(CC1)C1=CC=CC=C1

Specifications

Melting Point 5°C to 6°C
Density 0.945
Boiling Point 239°C to 240°C
Flash Point 98°C (208°F)
Odor Mild
Refractive Index 1.526
Quantity 500 g
UN Number UN3082
Beilstein 1906803
Solubility Information Immiscible with water.
Formula Weight 160.26
Percent Purity ≥97%
Chemical Name or Material Cyclohexylbenzene
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 212-572-0
RTECSNumber CZ1330000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only