missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylacetylene, 98%

Catalog No. p-7035694
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5341806 5 g
AAH5341814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5341806 Supplier Thermo Scientific Chemicals Supplier No. H5341806
Only null left

CAS: 931-48-6 | C8H12 | 108.184 g/mol

Cyclohexylacetylene is used in the preparation of hydrido-vinylidene complexes of osmium, which finds vital intermediates in several homogeneous and heterogeneous catalytic reactions such as alkene olimerization, polymerization and Fischer-Tropsch synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexylacetylene is used in the preparation of hydrido-vinylidene complexes of osmium, which finds vital intermediates in several homogeneous and heterogeneous catalytic reactions such as alkene olimerization, polymerization and Fischer-Tropsch synthesis.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 931-48-6
Molecular Formula C8H12
Molecular Weight (g/mol) 108.184
MDL Number MFCD00001513
InChI Key SSDZYLQUYMOSAK-UHFFFAOYSA-N
Synonym cyclohexylacetylene, cyclohexylethyne, cyclohexane, ethynyl, cyclohexyl acetylene, ethynyl-cyclohexane, cyclohexane, ethynyl-6ci, 7ci, 8ci, 9ci
PubChem CID 70263
IUPAC Name ethynylcyclohexane
SMILES C#CC1CCCCC1

Specifications

Density 0.828
Boiling Point 130°C to 132°C
Flash Point 18°C (64°F)
Refractive Index 1.454
Quantity 5 g
UN Number UN3295
Beilstein 1815535
Solubility Information Immiscible with water.
Formula Weight 108.18
Percent Purity 98%
Chemical Name or Material Cyclohexylacetylene
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 213-236-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only