accessibility menu, dialog, popup

UserName

CAS RN 931-48-6

IUPAC Name: ethynylcyclohexane
Synonyms: ethynylcyclohexane
Molecular Weight (g/mol): 108.18
Molecular Formula: C8H12
InChi Key: SSDZYLQUYMOSAK-UHFFFAOYSA-N
SMILES: C#CC1CCCCC1
Boiling Point 
  • (4)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)