missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexylacetic Acid 98.0+%, TCI America™
$163.64 - $847.08
Chemical Identifiers
| CAS | 5292-21-7 |
|---|---|
| Molecular Formula | C8H14O2 |
| Molecular Weight (g/mol) | 142.20 |
| MDL Number | MFCD00001518 |
| InChI Key | LJOODBDWMQKMFB-UHFFFAOYSA-N |
| Synonym | cyclohexylacetic acid, cyclohexaneacetic acid, cyclohexylethanoic acid, hexahydrophenylacetic acid, cyclohexyl acetic acid, unii-v43k3to0hn, fema no. 2347, cyclohexane-acetic acid, v43k3to0hn, cyclo-hexylacetic acid |
| PubChem CID | 21363 |
| ChEBI | CHEBI:37277 |
| IUPAC Name | 2-cyclohexylacetic acid |
| SMILES | OC(=O)CC1CCCCC1 |
Chemical Identifiers
| 5292-21-7 | |
| 142.20 | |
| LJOODBDWMQKMFB-UHFFFAOYSA-N | |
| 21363 | |
| 2-cyclohexylacetic acid |
| C8H14O2 | |
| MFCD00001518 | |
| cyclohexylacetic acid, cyclohexaneacetic acid, cyclohexylethanoic acid, hexahydrophenylacetic acid, cyclohexyl acetic acid, unii-v43k3to0hn, fema no. 2347, cyclohexane-acetic acid, v43k3to0hn, cyclo-hexylacetic acid | |
| CHEBI:37277 | |
| OC(=O)CC1CCCCC1 |
Specifications
| 5292-21-7 | |
| C8H14O2 | |
| 25 g | |
| LJOODBDWMQKMFB-UHFFFAOYSA-N | |
| 2-cyclohexylacetic acid | |
| 21363 | |
| 142.20 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00001518 | |
| cyclohexylacetic acid, cyclohexaneacetic acid, cyclohexylethanoic acid, hexahydrophenylacetic acid, cyclohexyl acetic acid, unii-v43k3to0hn, fema no. 2347, cyclohexane-acetic acid, v43k3to0hn, cyclo-hexylacetic acid | |
| OC(=O)CC1CCCCC1 | |
| 142.20 | |
| CHEBI:37277 | |
| ≥98.0% (GC,T) | |
| Cyclohexylacetic Acid |
Safety and Handling
RTECSNumber : GU6490000
TSCA : Yes