missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexylacetic Acid 98.0+%, TCI America™
Supplier: TCI America C0589250G
Specifications
| Cyclohexylacetic Acid | |
| White-Yellow | |
| MFCD00001518 | |
| cyclohexylacetic acid, cyclohexaneacetic acid, cyclohexylethanoic acid, hexahydrophenylacetic acid, cyclohexyl acetic acid, unii-v43k3to0hn, fema no. 2347, cyclohexane-acetic acid, v43k3to0hn, cyclo-hexylacetic acid | |
| OC(=O)CC1CCCCC1 | |
| 142.20 | |
| CHEBI:37277 | |
| ≥98.0% (GC,T) |
| 5292-21-7 | |
| C8H14O2 | |
| 250 g | |
| LJOODBDWMQKMFB-UHFFFAOYSA-N | |
| 2-cyclohexylacetic acid | |
| 21363 | |
| 142.20 | |
| Crystalline Powder |
Chemical Identifiers
| 5292-21-7 | |
| 142.20 | |
| LJOODBDWMQKMFB-UHFFFAOYSA-N | |
| 21363 | |
| 2-cyclohexylacetic acid |
| C8H14O2 | |
| MFCD00001518 | |
| cyclohexylacetic acid, cyclohexaneacetic acid, cyclohexylethanoic acid, hexahydrophenylacetic acid, cyclohexyl acetic acid, unii-v43k3to0hn, fema no. 2347, cyclohexane-acetic acid, v43k3to0hn, cyclo-hexylacetic acid | |
| CHEBI:37277 | |
| OC(=O)CC1CCCCC1 |
Safety and Handling
RTECSNumber : GU6490000
TSCA : Yes